Learn More
Medchemexpress LLC YNT-185 dihydrochloride | 1804978-82-2 | 99.8% | 688.66 g/mol | C33H39Cl2N5O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1361815MG
Encompass_Preferred
YNT-185 dihydrochloride is a nonpeptide, selective orexin type-2 receptor (OX2R) agonist supplied as the dihydrochloride salt for research use. It shows potent activity at OX2R with selectivity over OX1R and is intended for in vitro and in vivo pharmacology studies.
  • Potent orexin type-2 receptor agonist (EC50 = 28 nM at human OX2; OX1 EC50 = 2.75 μM).
  • High chemical purity: 99.78%.
  • Characterized compound with CAS 1804978-82-2 and formula C33H39Cl2N5O5S.
  • Provided as the dihydrochloride salt to aid handling and solubility.
  • Available in milligram quantities and as a 10 mM solution in DMSO for convenience.
  • Suitable for in vitro and in vivo orexin receptor pharmacology research.
Catalog No. 50-002-22549