Learn More
TARGETMOL CHEMICALS INC YODA 1 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T750610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Yoda1 is a Piezo1 ion channel agonist (EC50s = 17.1 and 26.6 uM for murine and human Piezo1-transfected HEK293T cells respectively)purity: 99%
Catalog No. 50-257-6310