Learn More
Medchemexpress LLC HY-18723 5mg Medchemexpress, Yoda 1 CAS:448947-81-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY187235MG
Encompass_Preferred
Medchemexpress, HY-18723 5mg Yoda 1 CAS:448947-81-7 Yoda 1 is a Piezo1 agonist. Yoda 1 activates purified Piezo1 channels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2078