Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Yohimbine-13C,d3 | 1261254-59-4 | 99.0% | 1 MG
SDP

Supplier:  Medchemexpress LLC HY12715S1MG

Encompass_Preferred

Yohimbine-13C,d3 is a 13C- and deuterium labeled version of Yohimbine, known as a potent and relatively nonselective alpha 2-adrenergic receptor (AR) antagonist with an IC50 of 0.6 μM. This stable isotope-labeled compound appears as a white to off-white solid and is ideal for use as a tracer or internal standard in quantitative analytical methods such as NMR, GC-MS, or LC-MS. It boasts a high purity of 99.0%. For optimal stability, store as a powder at -20°C for up to 3 years or 4°C for 2 years, or in solvent at -80°C for 6 months or -20°C for 1 month.

  • Stable isotope-labeled compound
  • Potent alpha 2-adrenergic receptor antagonist
  • Suitable for use as a tracer
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, and LC-MS methods
  • Appears as a white to off-white solid
  • Purity of 99.0%
  • Specific storage conditions for powder and solvent forms

Catalog No. 50-003-42418


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.