Learn More
Abcam Yohimbine hydrochloride, alpha2 adrenoceptor antagonist, 1G
Supplier:  Abcam AB1202391G
Encompass_Preferred
MW 390.9 Da, Purity >98%. Selective α2 adrenoceptor antagonist (pKi values are 8.52, 8.00 and 9.17 for human α2A, α2B and α2C receptors, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7762