Learn More
Medchemexpress LLC Yonkenafil hydrochloride | 804519-64-0 | 99.4% | 524.08 g·mol⁻¹ | C24H34ClN5O4S | 10MM 1ML

Supplier: Medchemexpress LLC HY12509510MM 1ML
Yonkenafil hydrochloride is a research-grade phosphodiesterase-5 (PDE5) inhibitor supplied as a high-purity hydrochloride salt for preclinical pharmacology and biochemical assays. It is available as a ready-to-use 10 mM DMSO solution or as a solid for reconstitution.
- High purity suitable for research use.
- Provided as 10 mM solution in DMSO or solid for reconstitution.
- Molecular weight 524.08 g·mol⁻¹; chemical formula C24H34ClN5O4S.
- Soluble in DMSO (≈190.8 mM); ultrasonic assistance may be required.
- Solid storage at 4°C; solutions stable at -80°C (up to 6 months) or -20°C (up to 1 month).
- Intended for preclinical studies involving PDE5 modulation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.