Learn More
Bioworld YPD agar base, 250 g
Supplier:  Bioworld 306201021
Encompass_Preferred
YPD Base is used with added carbohydrates for maintaining and propagating yeasts in molecular biology applications. Th addition of protein and yeast extract promotes better growth. Peptone 20 g/l Yeast Extract 10 g/l Agar 15 g/l This product is sold for laboratory research use only.
Catalog No. 50-254-400