Learn More
Bioworld YPD Broth, 500 g
Supplier:  Bioworld 306260171
Encompass_Preferred
YPD BrothFor propagation and maintenance of Saccharomyces cerevisiae and other yeast species for molecular studiesDehydrated Appearance: beige, homogeneous powderPrepared Appearance: light to medium amber brothSoluble in waterContains :Dextrose: 20g/LBacto Peptone: 20g/LYeast Extract: 10g/L Final pH (@25°C): 6.5 ±0.2Direction to use: Suspend 50g of powdered media per liter of distilled water. Sterilize by autoclaving at 121°C for 15 minutes.
Catalog No. 50-254-489