Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp YPD Broth 1G Capsules 1 KG
SDP

Supplier:  Research Products International Corp Y200911000.0

Encompass_Preferred

Add 50 capsules to 1 liter distilled water and heat until completely dissolved. Sterilize by autoclaving at 121°C for 15 minutes.

Ingredients:

Peptone 20 g/l
Dextrose 20 g/l
Yeast Extract 10 g/l

Catalog No. 50-223-0471


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.