Learn More
Alkali Scientific YPD Broth, 500 mL Capsules, 500 Grams
SDP
Supplier:  Alkali Scientific YPD001CP
Encompass
YPD Broth is a standard growth medium used for yeast culture. Available in 500g of powdered capsules (500mL per capsule), this medium provides a rich source of nutrients essential for supporting the growth of Saccharomyces cerevisiae and other yeast species. The broth contains a combination of peptone, yeast extract, and dextrose, which supports rapid growth and cell division. Ideal for research in molecular biology and genetics, YPD Broth is commonly used in yeast transformation studies, fermentation research, and the maintenance of yeast strains. The pre-measured capsules offer a convenient, efficient method of preparing high-quality culture medium for laboratory use.
Catalog No. 50-006-61684