Learn More
Medchemexpress LLC YQA14 25mg | 1221408-42-9 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11618125MG
Encompass_Preferred
YQA14 is a selective dopamine D3 receptor antagonist investigated as an anti-addiction agent. In animal models it inhibits morphine- and cocaine-induced conditioned place preference (CPP). Supplied as a solid research chemical for in vitro and in vivo pharmacology studies; not for human or veterinary use.
  • High affinity for dopamine D3 receptor, useful for receptor pharmacology.
  • Demonstrated inhibition of morphine- and cocaine-induced CPP in animals.
  • Available in small pack sizes suitable for dose-ranging and animal studies.
  • Supplied as a solid powder with reported high purity for research use.
  • SMILES provided for cheminformatics and structural confirmation.
Catalog No. 50-002-29500