Learn More
Cayman Chemical YS-035hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1770510 MG
Encompass_Preferred
A verapamil derivative that dose-dependently blocks calcium uptake via Na+/Ca2+ exchange in diverse types of cells (Ki = 28 µM); also prevents calcium release from mitochondria induced by Ruthenium Red but not by uncouplers, respiratory inhibitors, or a chelating agent
Catalog No. 50-296-8183