Learn More
Medchemexpress LLC HY-U00137 1mg Medchemexpress, YS-201 CAS:108852-42-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU001371MG
Encompass_Preferred
Medchemexpress, HY-U00137 1mg YS-201 CAS:108852-42-2 YS-201 is a dihydropyridine-type calcium channel antagonist previously in clinical trials for the treatment of angina pectoris and hypertension. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4424