Learn More
ABclonal Technology YTHDF1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1326020UL
Encompass_Preferred
Enables N6-methyladenosine-containing RNA binding activity and ribosome binding activity. Involved in mRNA destabilization, positive regulation of translational initiation, and stress granule assembly. Located in P-body and cytoplasmic stress granule.
Catalog No. 50-000-97746