Learn More
Medchemexpress LLC Ythdf2-in-1 | 99.4% | 358.35 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY173008100MG
Encompass_Preferred
YTHDF2-IN-1 (Compound CK-75) is an inhibitor for YT521-B homology domain family 2 YTHDF2 (Kd=26.2 μM), which blocks the interaction between YTHDF2 and m6A RNA.
  • It inhibits the colony formation of JAR cells.
  • It exhibits antiproliferative activity in various cancer cells (IC50 in micromolar levels).
  • It arrests the cell cycle at G0/G1 phase.
  • It induces apoptosis in K562.
Catalog No. 50-003-85625