Learn More
SPEX CERTIPREP LLC 3P YTTRIUM -100UG/ML-2%HNO3,100ML
Supplier:  SPEX CERTIPREP LLC 3P 100671100
Encompass_Preferred
(HPS by Zeptometrix) Yttrium at 100 g/mL in 2% HNO3 Yttrium (Y) ICP Single-Component Standard at 100 g/mL in 2% HNO3.This standard is traceable to NIST SRM 3167. ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited.
Catalog No. NC2243586