Learn More
Strem, An Ascensus Company CAS# 207801-29-4. 25g. Yttrium(III) acetylacetonate hydrate (99.9%-Y) (REO). MFCD00216639
SDP
Supplier:  Strem, An Ascensus Company 93390125G
Encompass
CAS# 207801-29-4. 25g. Yttrium(III) acetylacetonate hydrate (99.9%-Y) (REO). MFCD00216639. Molecular Weight: 386.24. Molecular Formula: Y(CH3COCHCOCH3)3∙XH2O. Color/form: white to yellow xtl. Strem# 93-3901. http://www.strem.com/catalog/v/93-3901/
Catalog No. 50-901-15201