Learn More
Selleck Chemical LLC YUM70
SDP
Supplier:  Selleck Chemical LLC S61935MG
Encompass_Preferred
YUM70 is a potent inhibitor of glucose-regulated protein 78 (GRP78) inhibitor with an IC50 of 1 5 M YUM70 induces endoplasmic reticulum (ER) stress-mediated apoptosis in pancreatic cancer Although YUM70 inhibits GRP78 enzymatic activity it increases the expression of GRP78 by increasing the chaperone translation mechanism
Catalog No. 50-217-3177