Learn More
ABclonal Technology YWHAZ Rabbit pAb
SDP
Supplier:  ABclonal Technology A133701000UL
Encompass_Preferred
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and sheep orthologs. The encoded protein interacts with IRS1 protein, suggesting a role in regulating insulin sensitivity. Several transcript variants that differ in the 5 UTR but that encode the same protein have been identified for this gene.
Catalog No. 50-000-97556