Learn More
Medchemexpress LLC Yx968 | 2742732-92-7 | 98.06% | 831.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16423350MG
Encompass_Preferred
YX968 is a potent and selective HDAC3/8 PROTAC dual degrader that exhibits antitumor activity by promoting apoptosis. It consists of a target protein ligand, a linker, and an E3 ligase ligand.
  • Induces cell apoptosis by blocking G1 phase
  • Cleavage of CASP3 was induced in a dose-dependent manner
  • Targets HDAC3 with a DC50 of 1.7 nM
  • Targets HDAC8 with a DC50 of 6.8 nM
Catalog No. 50-003-94652