Learn More
Medchemexpress LLC YZ9 | 6093-71-6 | 98.9% | 234.21 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11015610MG
Encompass_Preferred
YZ9 is a potent PFKFB3 inhibitor with an IC50 of 0.183 μM. It functions as a competitive inhibitor against Fru-6-P with a Ki value of 0.094 μM. YZ9 has demonstrated antitumor effects against cervical cancer.
  • Potent PFKFB3 inhibitor (IC50 of 0.183 μM)
  • Competitive inhibitor against Fru-6-P (Ki value of 0.094 μM)
  • Exhibits antitumor effects against cervical cancer
  • Purity: 98.89%
Catalog No. 50-003-44320