Learn More
Medchemexpress LLC YZ9 | 6093-71-6 | 98.9% | 234.21 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY110156100MG
Encompass_Preferred
YZ9 is a potent PFKFB3 inhibitor with an IC50 of 0.183 μM. It acts as a competitive inhibitor against Fru-6-P with a Ki value of 0.094 μM. This compound exhibits antitumor effects against cervical cancer, demonstrating antiproliferative activity against human HeLa cells.
  • Potent PFKFB3 inhibitor
  • Exhibits antitumor effects against cervical cancer
  • Demonstrates antiproliferative activity against human HeLa cells (GI50: 2.7 μM)
  • Provided as a solid, with color ranging from light yellow to brown
  • Store powder at -20°C for 3 years
  • For research use only
Catalog No. 50-003-43320