Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-10-hydroxynortriptyline | 47132-19-4 | 99.7% | 279.38 g·mol⁻¹ | C19H21NO | 5 MG
SDP

Supplier:  Medchemexpress LLC HY100646A5MG

Encompass_Preferred

(Z)-10-Hydroxynortriptyline is a research-grade metabolite of nortriptyline supplied as a solid reference standard for analytical and biochemical studies. It is intended for laboratory research use only and not for human or veterinary applications.

  • CAS number: 47132-19-4.
  • Molecular formula: C19H21NO.
  • Molecular weight: 279.38 g·mol⁻¹.
  • Purity: 99.7%.
  • Physical form: white to off-white solid.
  • Available in small research pack sizes (e.g., 5 mg).
  • Supplied for laboratory research use only.

Catalog No. 50-002-14460


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.