Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-10-hydroxynortriptyline-d3 | 47132-19-4 | 99.0% | 282.40 g/mol | C19H18D3NO | 1 MG
SDP

Supplier:  Medchemexpress LLC HY100646AS1MG

Encompass_Preferred

(Z)-10-hydroxynortriptyline-d3 is the deuterium-labeled stable isotope of 10-hydroxynortriptyline, supplied as a high-purity analytical reference standard for research applications. It is intended for use as an internal standard, tracer, or reference material in LC-MS/MS and other mass spectrometry workflows.

  • Deuterium-labeled (d3) analogue of 10-hydroxynortriptyline.
  • Chemical formula C19H18D3NO.
  • Molecular weight 282.40 g/mol.
  • Purity 99.0%.
  • Supplied as a 1 mg vial for analytical use.
  • Suitable as an internal standard for LC-MS/MS quantitation.
  • Includes datasheet and certificate of analysis for verification.

Catalog No. 50-002-13862


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.