Learn More
Medchemexpress LLC (Z)-3-ferrocenylmethylidene-1,3-dihydro-2H-indol-2-one | 1137967-28-2 | MFCD18252658 | 99.6% | 329.17 g·mol⁻1 | C19H15FeNO | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1084445MG
Encompass_Preferred
(Z)-FeCP-oxindole is a small-molecule inhibitor that selectively targets vascular endothelial growth factor receptor 2 (VEGFR2). It inhibits VEGFR2 with an IC50 of 200 nM, shows additional activity against VEGFR1 and PDGFR isoforms at higher concentrations, and has demonstrated anticancer activity in cellular models. The compound is supplied as a solid powder and as a DMSO stock solution for research use.
  • Selective VEGFR2 inhibitor with IC50 of 200 nM.
  • Activity against VEGFR1 and PDGFRα/β at higher concentrations.
  • Demonstrated anticancer activity in cellular models.
  • Supplied as solid powder and as DMSO stock solution.
  • High purity (≈99.6%) suitable for research applications.
  • Stable when stored under recommended temperature conditions.
Catalog No. 50-002-63721