Learn More
Medchemexpress LLC Z-321 | 130849-58-0 | 99.7% | 344.47 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY191235MG
Encompass_Preferred
Z-321 is a prolylendopeptidase (PEP) inhibitor. It has been shown to decrease the mean lordosis quotient (LQ) in treated groups. Specifically, a dose of 300 mg/kg Z-321 was effective in inhibiting lordosis behavior without disturbing locomotor activity in female rats.
  • Inhibitor of prolylendopeptidase (PEP).
  • Decreases sexual receptivity (mean lordosis quotient).
  • Inhibits lordosis behavior without disturbing locomotor activity at 300 mg/kg.
Catalog No. 50-003-85833