Learn More
Medchemexpress LLC Z-3578 | 473687-20-6 | 98.72% | 467.37 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY172458
Encompass_Preferred
Z-3578 is an orally active small-molecule antagonist of MrgX2 with potent antipseudoallergic activity. It effectively inhibits mast cell degranulation and can be used for the study of pseudoallergic reactions.
  • Inhibits mast cell degranulation induced by substance P and C48/80
  • Suppresses the release of β-hexosaminidase
  • Significantly reduces the release of histamine and TNF-α
  • Decreases intracellular calcium flux
  • Alleviates paw swelling and dye extravasation in a mouse pseudoallergy model
  • Reduces serum histamine levels in a mouse pseudoallergy model
Catalog No. 50-003-83935