Learn More
Medchemexpress LLC (Z)-4-hydroxy tamoxifen-d5 | 164365-20-2 | MFCD07369456 | 98.3% | 392.54 | C26H24D5NO2 | 5 MG

Supplier: Medchemexpress LLC HY132477S5MG
(Z)-4-Hydroxy Tamoxifen-d5 is a deuterium-labeled internal standard of (Z)-4-Hydroxy Tamoxifen supplied as a high-purity solid for analytical and pharmacokinetic research. It is intended for use as a tracer or reference in LC-MS/MS and related assays and is for research use only, not for human use.
- High-purity deuterium-labeled internal standard (98.3% purity).
- Molecular weight 392.54.
- Chemical formula C26H24D5NO2.
- Solid powder form for analytical use.
- Available in small pack sizes (1 mg to 100 mg).
- Stable when stored as powder: -20°C (3 years) or 4°C (2 years).
- Suitable for LC-MS/MS, pharmacokinetic, and tracer studies; research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.