Learn More
Abcam (Z)-4-Hydro x ytamo x ifen, estrogen receptor modulator, 10MG
Supplier:  Abcam AB14194310MG
Encompass_Preferred
MW 387.5 Da, Purity >98%. Potent and selective estrogen receptor antagonist (IC₅₀ = 7 nM). Active metabolite of (Z)-tamoxifen (ab120656) and exhibits higher potency in vitro. Shows antiproliferative effect (IC₅₀ = 27 μM). Induces p21 nuclear localization.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3338