Learn More
Medchemexpress LLC (Z)-4EGI-1 | 901787-88-0 | 98.0% | 451.28 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY19831A10MM/1ML
Encompass_Preferred
(Z)-4EGI-1 is the Z-isomer of 4EGI-1 and acts as an inhibitor of eIF4E/eIF4G interaction and translation initiation. It binds to eIF4E with an IC50 of 43.5 μM and a Kd value of 8.74 μM. This compound also exhibits anticancer activity.
  • Inhibits eIF4E/eIF4G interaction and translation initiation.
  • Effectively binds to eIF4E.
  • Has anticancer activity.
  • Reduces expressions of regulatory proteins like cyclin D1, cyclin E, and Survivin in CRL-2813 melanoma cells.
  • Inhibits cancer cell proliferation in CRL-2351 breast cells and CRL-2813 melanoma cells.
Catalog No. 50-004-01897