Learn More
Medchemexpress LLC (Z)-5-(4-Iodobenzylidene)-2-thioxothiazolidin-4-one | 181765-52-6 | 98.0% | 347.20 | 10 G

Supplier: Medchemexpress LLC HY2193710G
(Z)-5-(4-Iodobenzylidene)-2-thioxothiazolidin-4-one is a chemical compound used as a drug intermediate for the synthesis of various active compounds. It is intended for research use only and is not sold to patients.
- Molecular weight of 347.20
- Chemical formula is C10H6INOS2
- CAS number 181765-52-6
- Appears as a solid
- Color is light yellow to yellow
- Purity of 98.0%
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.