Learn More
Medchemexpress LLC Z -8-Dodecen-1-ol 10mg
SDP
Supplier:  Medchemexpress LLC HYW10841810MG
Encompass_Preferred
(Z)-8-Dodecen-1-ol (cis-8-Dodecen-1-ol) is an active straight chain lepidopteran pheromone (SCLP)[1]
Catalog No. 50-002-05679