Learn More
Medchemexpress LLC (Z)-9-propenyladenine | 1464851-21-5 | MFCD20705512 | 98.0% | 175.19 g/mol | C8H9N5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100079A10MM 1ML
Encompass_Preferred
(Z)-9-Propenyladenine is the Z-isomer of a mutagenic impurity found in tenofovir disoproxil fumarate, provided as a ready-to-use 10 mM solution in DMSO (1 mL) for analytical reference and impurity profiling. The compound has formula C8H9N5, CAS 1464851-21-5, molecular weight 175.19 g/mol, and listed purity 98.02%.
  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as a 1 mL aliquot for single-use analytical assays.
  • High purity suitable for reference and method development.
  • Characterized by CAS 1464851-21-5 and formula C8H9N5.
  • Appropriate for impurity profiling of tenofovir disoproxil-related products.
Catalog No. 50-002-15364