Learn More
Medchemexpress LLC (Z)-9-propenyladenine | 1464851-21-5 | MFCD30718180 | 97.0% | 175.19 g/mol | C8H9N5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100079A10MG
Encompass_Preferred
(Z)-9-Propenyladenine is the Z-isomer impurity of tenofovir disoproxil fumarate, provided as an analytical standard for impurity profiling, method development, and analytical testing. Available as solid material in multiple milligram sizes or as a DMSO solution; intended for research use only.
  • Analytical standard for impurity identification and quantification.
  • Available as solid and solution formats to suit assay needs.
  • Supporting documentation available: data sheet, COA, LCMS, and SDS.
  • Storage recommendations provided to retain stability and integrity.
  • Suitable for analytical method development and quality control workflows.
Catalog No. 50-002-15710