Learn More
Medchemexpress LLC (Z)-9-propenyladenine | 1464851-21-5 | MFCD30718180 | 98.0% | 175.19 g/mol | C8H9N5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100079A50MG
Encompass_Preferred
(Z)-9-propenyladenine is the Z-isomer mutagenic impurity of tenofovir disoproxil fumarate supplied as an analytical standard for research and analytical applications. It is intended for use as a reference material in qualitative and quantitative analyses, and in method development and validation for chromatographic and mass spectrometry techniques.
  • Analytical standard grade for accurate quantitation.
  • Suitable for HPLC, GC, and MS method development.
  • High purity (~98.0%) with certificate of analysis and SDS available.
  • Available in multiple solid pack sizes, including a 50 mg option.
  • Molecular weight 175.19 g/mol; molecular formula C8H9N5.
Catalog No. 50-002-15204