Learn More
Medchemexpress LLC (Z)-9-Tricosene (cis-9-Tricosene) | 27519-02-4 | 98.0% | 322.61 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW009165100MG
Encompass_Preferred
(Z)-9-Tricosene (cis-9-Tricosene) is an insect pheromone present in flies of the order Diptera (such as the housefly) and can be used as an insecticide.
  • Functions as an insect pheromone, specifically present in Diptera flies.
  • Can be utilized as an insecticide.
Catalog No. 50-004-53505