Learn More
Medchemexpress LLC Z-aevd-fmk | 1135688-47-9 | 95.0% | 610.63 | C28H39FN4O10 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12530550MG
Encompass_Preferred
Z-AEVD-FMK is a peptide-based caspase-10 inhibitor used in cell biology research to block caspase-10-mediated apoptotic signaling. It inhibits activation of Bid and the mitochondrial release of apoptosis-inducing factor (AIF), resulting in reduced apoptotic cell counts in treated cultures. The compound is supplied as a solid research reagent and should be stored protected from light and under inert atmosphere; solution storage recommendations vary by solvent.
Catalog No. 50-002-37953