Learn More
Medchemexpress LLC (Z-Ala-Ala-Ala-Ala)2Rh110 | 149695-85-2 | 97.2% | 1167.22 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP31061MG
Encompass_Preferred
(Z-Ala-Ala-Ala-Ala)2Rh110 is a sensitive fluorogenic elastase substrate. This colorless and nonfluorescent compound is selectively cleaved by elastase to yield the highly fluorescent rhodamine 110, which can be analyzed with an excitation wavelength of 485 nm and emission wavelength of 525 nm. It has a chemical formula of C60H66N10O15.
  • Sensitive fluorogenic elastase substrate
  • Selectively cleaved by elastase
  • Yields highly fluorescent rhodamine 110
  • Appearance: solid
  • Color: light yellow to yellow
Catalog No. 50-003-99437