Learn More
Medchemexpress LLC Z-Ala-Ala-Asn-AMC (Cbz-Ala-Ala-Asn-AMC) | 149697-16-5 | MFCD02684274 | 99.4% | 565.57 | C28H31N5O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1366265MG
Encompass_Preferred
Z-Ala-Ala-Asn-AMC (Cbz-Ala-Ala-Asn-AMC) is a synthetic fluorogenic peptide substrate for the protease legumain. Cleavage of the peptide releases 7-amino-4-methylcoumarin (AMC), enabling quantitative fluorometric measurement of legumain activity in biochemical and cell-based assays.
Catalog No. 50-002-22040