Learn More
Carbosynth LLC. Z-Ala-Ala-Asn-MCA (5 mg vial), 1ea, MW: 565.57, Molecular formula: C28H31N5O8, CAS# 149697-16-5
SDP
Supplier:  Carbosynth LLC. MCA3209V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Benzyloxycarbonyl-L-Alanyl-L-Alanyl-L-Asparagine-4-Methyl-Coumaryl-7-Amide, Z-Ala-Ala-Asn-AMC, Z-AAN-AMC, ZAAN-MCA, Application: Substrate for Legumain, Storage Temperature: -20C, References: A.A. Kembhavi, D.J. Buttle, C.G. Knight, and A.J. Barret, Arch. Biochem. Biophys., 303, 208 (1993)., J.-M. Chen, P.M. Dando, N.D. Rawlings, M.A. Brown, N.E. Young, R.A. Stevens, E. Hewitt, C. Watts, and A.J. Barret, J. Biol. Chem., 272, 8090 (1997)., B. Manoury, E.W. Hewitt, N. Morrice, P.M. Dando, A.J. Barret, and C. Watts, Nature, 396, 695 (1998).
Catalog No. 50-168-5950