Learn More
Medchemexpress LLC Z-Ala-Ala-Asp-CMK | 183284-21-1 | 98.92% | 441.86 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1000410MG
Encompass_Preferred
Z-Ala-Ala-Asp-CMK (Z-AAD-CMK) is a selective granzyme B inhibitor. It functions by binding to the active site of granzyme B, thereby blocking its proteolytic activity. This compound exhibits anti-inflammatory properties and is utilized in research related to inflammatory diseases and cancer.
  • Selective granzyme B inhibitor
  • Blocks proteolytic function of granzyme B
  • Exhibits anti-inflammatory activity
  • Can be used in research for inflammatory diseases and cancer
  • Inhibits granzyme B-induced IL-18 activation and IFN-γ secretion
  • Reduces DNA fragmentation
  • Inhibits caspase-3 activity and ROS production
Catalog No. 50-004-25135