Learn More
Medchemexpress LLC Z-Ala-Ala-Asp-CMK | 183284-21-1 | 98.92% | 441.86 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP100045MG
Encompass_Preferred
Z-Ala-Ala-Asp-CMK (Z-AAD-CMK) is a selective granzyme B inhibitor. It functions by binding to the active site of granzyme B, thereby blocking its proteolytic function. This compound exhibits anti-inflammatory activity and is utilized in research for inflammatory diseases and cancer studies.
  • Selective granzyme B inhibitor
  • Blocks proteolytic function of granzyme B
  • Exhibits anti-inflammatory activity
  • Used in research for inflammatory diseases and cancer
  • Inhibits granzyme B-induced IL-18 activation
  • Reduces DNA fragmentation
  • Inhibits caspase-3 activity
  • Reduces ROS production
Catalog No. 50-004-25060