Learn More
Medchemexpress LLC Z-Ala-Glu(OMe)-Val-Asp(OMe)-fluoromethyl ketone | 1135688-47-9 | MFCD03453601 | ≥95.0% | 610.63 g·mol⁻¹ | C28H39FN4O10 | 1MG

Supplier: Medchemexpress LLC HY1253051MG
Z-AEVD-FMK is a cell-permeable, irreversible caspase-10 inhibitor used in apoptosis research. It is a fluoromethyl ketone-derivatized tetrapeptide that inhibits Bid activation and the release of apoptosis-inducing factor from mitochondria, reducing apoptotic cell numbers. The compound is supplied in milligram quantities for laboratory studies.
- Irreversible caspase-10 inhibitor for apoptosis pathway studies.
- Fluoromethyl ketone-derivatized tetrapeptide structure.
- Purity ≥95%.
- Supplied as a solid in milligram-scale quantities for research use.
- Storage recommended at -20°C; protect from light and store under inert gas.
- Molecular weight 610.63 g·mol⁻¹; formula C28H39FN4O10.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.