Learn More
Medchemexpress LLC Z-Arg-Leu-Arg-Gly-Gly-AMC | 167698-69-3 | 99.9% | 848.95 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP43881MG
Encompass_Preferred
Z-Arg-Leu-Arg-Gly-Gly-AMC is a peptide substrate for SARS-CoV PLpro (Papain-like protease). It is intended for research use only.
  • High purity: 99.93%
  • Specific substrate for SARS-CoV PLpro
Catalog No. 50-003-98799