Learn More
Medchemexpress LLC Z-Arg-Leu-Arg-Gly-Gly-AMC | 167698-69-3 | 99.93% | 848.95 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP43885MG
Encompass_Preferred
Z-Arg-Leu-Arg-Gly-Gly-AMC is a peptide substrate of SARS-CoV PLpro, designed for use in research applications.
  • Peptide substrate for SARS-CoV PLpro
  • Designed for research applications
  • Useful in viral infection studies
  • Supports SARS-CoV research
Catalog No. 50-003-96734