Learn More
Medchemexpress LLC Z-Arg-Leu-Arg-Gly-Gly-AMC acetate | 98.85% | 848.95 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP4388A1MG
Encompass_Preferred
Z-Arg-Leu-Arg-Gly-Gly-AMC acetate is a peptide substrate for SARS-CoV PLpro.
  • Solid physical form
  • White to off-white color
  • Soluble in water (25 mg/mL with ultrasonication)
Catalog No. 50-003-96747