Learn More
Medchemexpress LLC Z-Asp-CH2-DCB | 153088-73-4 | 99.6% | 454.26 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11395310MM/1ML
Encompass_Preferred
Z-Asp-CH2-DCB is an irreversible broad-spectrum caspase inhibitor that also inhibits proteases with caspase-like activity. It blocks the production of several inflammatory cytokines and reduces T-cell proliferation in stimulated peripheral blood mononuclear cells.
  • Irreversible broad-spectrum caspase inhibitor
  • Inhibits proteases with caspase-like activity
  • Blocks production of IL-1β, TNF-α, IL-6, and IFN-γ
  • Reduces T-cell proliferation
  • Prevents SU5416-induced septal cell apoptosis and emphysema development
Catalog No. 50-004-36171