Learn More
Medchemexpress LLC Z-Asp-OBzl | 4779-31-1 | 99.5% | 357.36 | 100 G

Supplier: Medchemexpress LLC HYW008685100G
Z-Asp-OBzl is an aspartic acid derivative intended for research use only. Amino acids and their derivatives, like Z-Asp-OBzl, are recognized as beneficial ergogenic dietary substances, influencing anabolic hormone secretion, supplying fuel during exercise, improving mental performance, and preventing exercise-induced muscle damage.
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Powder at -20°C for 3 years, 4°C for 2 years. In solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.