Learn More
Medchemexpress LLC (Z)-Capsaicin | 25775-90-0 | 99.8% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1583
Encompass_Preferred
(Z)-Capsaicin is the cis isomer of capsaicin. It acts as an orally active TRPV1 agonist and is used in the research of neuropathic pain.
  • Acts as a TRPV1 agonist
  • Used in the research of neuropathic pain
  • Exhibits agonist activity at human TRPV1 expressed in CHO cells (EC50 of 28.2 nM)
  • May also block neuronal calcium channels
  • Significantly attenuates nociceptive behavior after oral administration in rats
  • Tolerated in experimental genital herpes simplex virus infection in guinea pigs
Catalog No. 50-004-14176