Learn More
TARGETMOL CHEMICALS INC Z-Capsaicin 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T110950MG
Encompass_Preferred
Also available in 10 mg and bulk. Please contact Fisher for quotes. (Z)-Capsaicin (Zucapsaicin) is a medication used to treat osteoarthritis of the knee and Others neuropathic pain.(Z)-Capsaicin is a synthetic cis isomer of natural capsaicin that has shown therapeutic efficacy in pain accompanying osteoarthritis of the knee. Purity 99.67%
Catalog No. 50-288-0609